Anti-ANXA2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310039EA 5425 NOK
ORIGTA310039
Anti-ANXA2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ANXA2 (annexin A2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR