Anti-BRCA1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310042EA 8500 NOK
ORIGTA310042
Anti-BRCA1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR