Anti-IGF1R Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310056EA 8500 NOK
ORIGTA310056
Anti-IGF1R Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR