Anti-KLK14 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310060EA 5625 NOK
ORIGTA310060
Anti-KLK14 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:100

Type: Primary
Antigen: KLK14 (kallikrein-related peptidase 14)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR