Anti-NEUROG3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310069EA 4600 NOK
ORIGTA310069
Anti-NEUROG3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: NEUROG3 (neurogenin 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR