Anti-ADAMTS5 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA310094EA 5425 NOK
ORIGTA310094
Anti-ADAMTS5 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ADAMTS5 (ADAM metallopeptidase with thrombospondin type 1 motif, 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR