Anti-FOXD3 Rabbit Polyclonal Antibody
Leverandør: OriGene
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000
Type: Primary
Antigen: FOXD3 (forkhead box D3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
BESTILL
Utvid alle / Skjul alle
Str Fp Art.nr Antall
100 µl 100 µl ORIGTA315355
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Antigen symbol FOXD3
Antigen name Forkhead Box D3
Reaktivitet Human, Mouse
Konjugasjon Unconjugated
Klonalitet Polyclonal
antistofftype Primary
Vert Rabbit
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Epitope internal
Kyssabsorpsjon none
Rensing purified by affinity chromatography
Form Liquid
Lagringsbuffer Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Lagringstemp Cool
Frakttemp. Ice
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall