Anti-R3HCC1L Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA315380EA 6500 NOK
ORIGTA315380
Anti-R3HCC1L Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: R3HCC1L (R3H domain and coiled-coil containing 1-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR