Anti-MRPL18 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA315391EA 6500 NOK
ORIGTA315391
Anti-MRPL18 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: MRPL18 (mitochondrial ribosomal protein L18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR