Anti-HUNK Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA315410EA 6500 NOK
ORIGTA315410
Anti-HUNK Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: HUNK (hormonally up-regulated Neu-associated kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR