Anti-TAS2R13 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA315430EA 6500 NOK
ORIGTA315430
Anti-TAS2R13 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: TAS2R13 (taste receptor, type 2, member 13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR