Anti-HSD17B11 Rabbit Polyclonal Antibody

ORIGTA315448EA 3700 NOK
ORIGTA315448
Anti-HSD17B11 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: HSD17B11 (hydroxysteroid (17-beta) dehydrogenase 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR