Anti-PAIP1 Mouse Polyclonal Antibody
Leverandør: Abnova
ABNOH00010605-A01EA 2975 NOK
ABNOH00010605-A01
Anti-PAIP1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human