Anti-KCNJ4 Mouse Monoclonal Antibody

Leverandør: Biorbyt
BIRBORB95885-50EA 0 NOK
BIRBORB95885-50
Anti-KCNJ4 Mouse Monoclonal Antibody
Antistoffer
Immunofluorescence: 0.1-1 ?g/ml; Immunochemistry: 5 ?g/ml; Immunoprecipitation: 1-10 ?g/ml; Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: KCNJ4 (potassium inwardly-rectifying channel, subfamily J, member 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR