Hold min sesjon åpen
Slutter 
Sesjonen har utløpt
Økten er utløpt. For sikkerhets skyld har vi logget deg ut. Vil du logge inn på nytt?
Prices for promotional code products are currently not displayed correctly on vwr.com – we are working on this issue

Anti-TRPC7 Mouse monoclonal antibody unconjugated

Leverandør: Biorbyt
Immunofluorescence: 0.1-1 μg/ml; Immunochemistry: 10 μg/ml; Immunoprecipitation: 1-10 μg/ml; Western Blot: 1-10 μg/ml

Type: Primary
Antigen: TRPC7 (transient receptor potential cation channel, subfamily C, member 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat



 

BESTILL

Utvid alle / Skjul alle
Str Fp Art.nr Antall
50 µg 50 µG BIRBORB95888-50
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog

SPESIFIKASJONER

Antigen symbol TRPC7
Antigen name transient receptor potential cation channel, subfamily C, member 7
Reaktivitet Rat, Mouse, Human
Konjugasjon Unconjugated
Klonalitet Monoclonal
antistofftype Primary
Vert Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Rensing purified by Protein G chromatography
Lagringsbuffer PBS, pH 7,4, 50% glycerol, 0,09% sodium azide.
Lagringstemp Cool
Frakttemp. Ice
Legg inn detaljer

Velg varen hvor du søker etter sertifikat

Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk

Skriv inn serifikatdetaljer

Skriv inn lot nummer:

Velg fra de siste batch / lot tallene:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall