Anti-EI24 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
BIRBORB95897-50EA 0 NOK
BIRBORB95897-50
Anti-EI24 Rabbit Polyclonal Antibody
Antistoffer
Immunochemistry: 5 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: EI24 (etoposide induced 2.4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR