Anti-GPR17 Goat Polyclonal Antibody

Leverandør: Biorbyt
BIRBORB95900-50EA 0 NOK
BIRBORB95900-50
Anti-GPR17 Goat Polyclonal Antibody
Antistoffer
ELISA: 1:1000; Immunochemistry: 10 ?g/ml; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: GPR17 (G protein-coupled receptor 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR