Anti-PLXNB1 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
BIRBORB95905-50EA 0 NOK
BIRBORB95905-50
Anti-PLXNB1 Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:10000; Immunochemistry: 5 ?g/ml; Western Blot: 1:200

Type: Primary
Antigen: PLXNB1 (plexin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR