Anti-CD28 Armenian Hamster Monoclonal Antibody (Biotin) [clone: PV-1]
Catalog # ABCAAB25178-250
Supplier: Abcam
Supplier Article Number: AB25178-250
Local Article Number: ABCAAB25178-250
Price
kr 7 775,00
Log in for availability
Specifications
- Antibody type:Primary
- Antigen name:CD28 molecule
- Antigen symbol:CD28
- Clonality:Monoclonal
- Clone:PV-1
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Armenian hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pk:250 µG