Anti-CD3E Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB27434-7EA 11175 NOK
ABCAAB27434-7
Anti-CD3E Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR