Prices for promotional code products are currently not displayed correctly on vwr.com – we are working on this issue
Anti-Melanoma gp100 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
Type: Primary
Antigen: Melanoma gp100
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
BESTILL
Utvid alle / Skjul alle
Str Fp Art.nr Antall
500 µl 500 µl ABCAAB27435-500
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Antigen name Melanoma gp100
Reaktivitet Human
Konjugasjon Unconjugated
Klonalitet Polyclonal
antistofftype Primary
Vert Rabbit
Isotype IgG
ImmunoChemistry Yes
Lagringstemp Cool
Frakttemp. Ice
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall