Anti-BIRC5 Goat Polyclonal Antibody

Leverandør: Abcam
ABCAAB27468-100EA 8425 NOK
ABCAAB27468-100
Anti-BIRC5 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR