Anti-TRIM62 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB51039-100EA 8350 NOK
ABCAAB51039-100
Anti-TRIM62 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TRIM62 (tripartite motif containing 62)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human