Anti-INPP5J Rabbit polyclonal antibody unconjugated

ABCAAB61147-50EA 7175 NOK
ABCAAB61147-50
Anti-INPP5J Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: INPP5J (inositol polyphosphate-5-phosphatase J)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR