Anti-ELK3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB61164-100EA 8500 NOK
ABCAAB61164-100
Anti-ELK3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: ELK3 (ELK3, ETS-domain protein (SRF accessory protein 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR