Anti-FNIP1 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB61395-50EA 11400 NOK
ABCAAB61395-50
Anti-FNIP1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FNIP1 (folliculin interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse