Anti-TATA binding protein TBP Mouse monoclonal antibody unconjugated [clone: 58C9]

ABCAAB61411-100EA 7725 NOK
ABCAAB61411-100
Anti-TATA binding protein TBP Mouse monoclonal antibody unconjugated [clone: 58C9]
Type: Primary
Antigen: TATA binding protein TBP
Clonality: monoclonal
Clone: 58C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR