Anti-FZD9 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB61430-50EA 11475 NOK
ABCAAB61430-50
Anti-FZD9 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FZD9 (frizzled class receptor 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat