Anti-KCNIP3 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB61770-100EA 11850 NOK
ABCAAB61770-100
Anti-KCNIP3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KCNIP3 (Kv channel interacting protein 3, calsenilin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR