Anti-PGR Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB61785-100EA 11475 NOK
ABCAAB61785-100
Anti-PGR Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PGR (progesterone receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human