Anti-Eph receptor B1 + Eph receptor B2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB61791-100EA 12100 NOK
ABCAAB61791-100
Anti-Eph receptor B1 + Eph receptor B2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Eph receptor B1 + Eph receptor B2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR