Anti-hnRNP C1 + C2 Rabbit polyclonal antibody unconjugated

ABCAAB61796-100EA 7725 NOK
ABCAAB61796-100
Anti-hnRNP C1 + C2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: hnRNP C1 + C2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR