Anti-SSB Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB61800-100EA 11725 NOK
ABCAAB61800-100
Anti-SSB Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SSB (Sjogren syndrome antigen B (autoantigen La))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR