Anti-PDE8A Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB61815-50EA 11475 NOK
ABCAAB61815-50
Anti-PDE8A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PDE8A (phosphodiesterase 8A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human