Anti-PDE8B Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB61817-50EA 10900 NOK
ABCAAB61817-50
Anti-PDE8B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PDE8B (phosphodiesterase 8B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human