Anti-SGK3 Rabbit polyclonal antibody unconjugated

ABCAAB61818-50EA 7550 NOK
ABCAAB61818-50
Anti-SGK3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: SGK3 (serum/glucocorticoid regulated kinase family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR