Anti-HSPA13 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: C92F3A-5]
Leverandør: Abcam
ABCAAB61907-100EA 11475 NOK
ABCAAB61907-100
Anti-HSPA13 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: C92F3A-5]
Antistoffer
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: C92F3A-5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestill nå