Anti-F8 Chicken polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB61910-100EA 8350 NOK
ABCAAB61910-100
Anti-F8 Chicken polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: F8 (coagulation factor VIII, procoagulant component)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR