Anti-F7 Chicken polyclonal antibody unconjugated

ABCAAB61963-100EA 7550 NOK
ABCAAB61963-100
Anti-F7 Chicken polyclonal antibody unconjugated
Type: Primary
Antigen: F7 (coagulation factor VII (serum prothrombin conversion accelerator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR