Anti-STIM1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62031-100EA 11525 NOK
ABCAAB62031-100
Anti-STIM1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: STIM1 (stromal interaction molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR