Anti-TRPV2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62046-50EA 8500 NOK
ABCAAB62046-50
Anti-TRPV2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TRPV2 (transient receptor potential cation channel, subfamily V, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR