Anti-WNT10A Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62051-50EA 11725 NOK
ABCAAB62051-50
Anti-WNT10A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: WNT10A (wingless-type MMTV integration site family, member 10A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR