Anti-BRCC3 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB62075-100EA 11725 NOK
ABCAAB62075-100
Anti-BRCC3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: BRCC3 (BRCA1/BRCA2-containing complex, subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat