Anti-CCND1 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB62151-100EA 11475 NOK
ABCAAB62151-100
Anti-CCND1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human