Anti-COX IV Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: mAbcam 62164]
Leverandør: Abcam
ABCAAB62164-100EA 12975 NOK
ABCAAB62164-100
Anti-COX IV Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: mAbcam 62164]
Antistoffer
Type: Primary
Antigen: COX IV
Clonality: monoclonal
Clone: mAbcam 62164
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human