Anti-SLC6A8 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62196-100EA 8425 NOK
ABCAAB62196-100
Anti-SLC6A8 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC6A8 (solute carrier family 6 (neurotransmitter transporter), member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR