Anti-HSD17B6 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62221-100EA 8550 NOK
ABCAAB62221-100
Anti-HSD17B6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSD17B6 (hydroxysteroid (17-beta) dehydrogenase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR