Anti-PTPN1 Rabbit Monoclonal Antibody [clone: EP1839Y]
Leverandør: Abcam
ABCAAB62330-100EA 10650 NOK
ABCAAB62330-100
Anti-PTPN1 Rabbit Monoclonal Antibody [clone: EP1839Y]
Antistoffer
Type: Primary
Antigen: PTPN1 (protein tyrosine phosphatase, non-receptor type 1)
Clonality: monoclonal
Clone: EP1839Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå