Vårt Oslo-kontor flytter den 25. mars 2019 til Brynsalleen 4, 0667 Oslo
Anti-RFP Rabbit Polyclonal Antibody
Leverandør: Abcam
Red fluorescent protein cloned from Discosoma coral (DsRed or drFP583) is an important tool in biotechnology and cell biology as a spectrally distinct companion or substitute for the green fluorescent protein (GFP; from Aequorea jellyfish).
Type: Primary
Antigen: RFP (Red Fluorescent Protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
BESTILL
Utvid alle / Skjul alle
Str Fp Art.nr Antall
100 µg 100 µG ABCAAB62341-100
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Antigen symbol RFP
Antigen name Red Fluorescent Protein
Konjugasjon Unconjugated
Klonalitet Polyclonal
antistofftype Primary
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Konsentrasjon 100 µg at 0,5 mg/ml
Immunogen Recombinant RFP expressed in E. coli
Rensing Immunogen affinity purified
Lagringsbuffer Preservative: 0,01% Thimerosal (merthiolate), Constituents: 30% Glycerol, 0,5% BSA, PBS, pH 7,2
Lagringstemp Cool
Frakttemp. Ice
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall