Anti-SYK Rabbit Monoclonal Antibody [clone: EP575Y]
Leverandør: Abcam
ABCAAB62350-100EA 11575 NOK
ABCAAB62350-100
Anti-SYK Rabbit Monoclonal Antibody [clone: EP575Y]
Antistoffer
Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: monoclonal
Clone: EP575Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå